Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GK SIRNA Assays ›
          Gene Symbol:
          GK GK3P
          Gene Name:
          glycerol kinase
          glycerol kinase 3 pseudogene
          Gene Aliases:
          GK1, GKD
          GKP3, GKTB
          Chromosome Location:
          Chr.X: 30653348 - 30731462 on Build GRCh38
          Chromosome Location:
          Chr.4: 165277792 - 165280023 on Build GRCh38
          UniGene ID:
          Hs.1466
          Hs.654557
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS178324
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000167.5 NP_000158.1 5 547
          NM_001128127.2 NP_001121599.1 5 547
          NM_001205019.1 NP_001191948.1 5 547
          NM_203391.3 NP_976325.1 5 547
          XM_005274488.4 XP_005274545.1 3 317
          XM_006724483.2 XP_006724546.1 6 584
          XM_006724484.2 XP_006724547.1 6 583
          XM_006724485.2 XP_006724548.1 4 392
          XM_006724486.3 XP_006724549.1 3 377
          XM_011545491.2 XP_011543793.1 6 585
          XM_011545492.1 XP_011543794.1 6 584
          XM_011545493.2 XP_011543795.1 4 267
          XM_011545494.2 XP_011543796.1 4 212
          XM_017029409.1 XP_016884898.1 6 582
          XM_017029410.1 XP_016884899.1 3 361
          XM_017029411.1 XP_016884900.1 4 394
          XM_017029412.1 XP_016884901.1 3 315
          GenBank mRNA AK294304.1 - 4 303
          AK294938.1 - 6 582
          AK308238.1 - 3 226
          AK313215.1 - 5 483
          BC037549.1 - 5 404 5213716
          BC042421.1 - 5 548 4820945
          BC071595.1 - 5 548 5276056
          BM906503.1 - 494
          BX647604.1 - 5 368
          L13943.1 - 5 434
          X68285.1 - 3 140
          Gene Symbol: GK3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026575.1 1 746
          GenBank mRNA AK225740.1 - 1 397
          AK292282.1 - 1 746
          BC066960.1 - 1 402

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol metabolic process
          triglyceride metabolic process
          response to cold
          phosphorylation
          regulation of fatty acid metabolic process
          triglyceride biosynthetic process
          glycerol catabolic process
          response to drug
          glucose homeostasis
          response to ethanol
          glycerol-3-phosphate biosynthetic process
          glycerol kinase activity
          protein binding
          ATP binding
          histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline