Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DISC1 SIRNA Assays ›
          Gene Symbol:
          DISC1 TSNAX-DISC1
          Gene Name:
          disrupted in schizophrenia 1
          TSNAX-DISC1 readthrough (NMD candidate)
          Gene Aliases:
          C1orf136, SCZD9
          -
          Chromosome Location:
          Chr.1: 231626815 - 232041272 on Build GRCh38
          Chromosome Location:
          Chr.1: 231528653 - 232041272 on Build GRCh38
          UniGene ID:
          Hs.13318
          Hs.13318
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS178359
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DISC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012957.1 NP_001012975.1 2 683
          NM_001012958.1 NP_001012976.1 2 683
          NM_001012959.1 NP_001012977.1 2 683
          NM_001164537.1 NP_001158009.1 2 683
          NM_001164538.1 NP_001158010.1 2 683
          NM_001164539.1 NP_001158011.1 2 683
          NM_001164540.1 NP_001158012.1 2 683
          NM_001164541.1 NP_001158013.1 2 683
          NM_001164542.1 NP_001158014.1 2 683
          NM_001164544.1 NP_001158016.1 2 683
          NM_001164545.1 NP_001158017.1 2 683
          NM_001164546.1 NP_001158018.1 2 683
          NM_001164547.1 NP_001158019.1 2 683
          NM_001164548.1 NP_001158020.1 2 683
          NM_001164549.1 NP_001158021.1 2 683
          NM_001164550.1 NP_001158022.1 2 683
          NM_001164551.1 NP_001158023.1 2 683
          NM_001164552.1 NP_001158024.1 2 683
          NM_001164553.1 NP_001158025.1 2 683
          NM_001164554.1 NP_001158026.1 2 683
          NM_001164555.1 NP_001158027.1 2 683
          NM_018662.2 NP_061132.2 2 683
          GenBank mRNA AB007926.1 - 2 669
          AF222980.1 - 2 683
          AJ506177.1 - 2 683
          AJ506178.1 - 2 683
          AK025293.1 - 2 748
          AK096457.1 - 3 855
          BC151225.1 - 2 669 9007180
          FJ804174.1 - 2 683
          FJ804175.1 - 2 683
          FJ804176.1 - 2 683
          FJ804177.1 - 2 683
          FJ804178.1 - 2 683
          FJ804179.1 - 2 683
          FJ804180.1 - 2 683
          FJ804181.1 - 2 683
          FJ804182.1 - 2 683
          FJ804183.1 - 2 683
          FJ804184.1 - 2 683
          FJ804185.1 - 2 683
          FJ804186.1 - 2 683
          FJ804187.1 - 2 683
          FJ804188.1 - 2 683
          FJ804189.1 - 2 683
          FJ804190.1 - 2 683
          FJ804191.1 - 2 683
          FJ804192.1 - 2 683
          FJ804193.1 - 2 683
          FJ804194.1 - 2 683
          FJ804195.1 - 3 751
          FJ804196.1 - 2 683
          FJ804197.1 - 2 683
          FJ804198.1 - 2 683
          FJ804199.1 - 2 683
          FJ804200.1 - 2 683
          FJ804201.1 - 2 683
          FJ804202.1 - 2 683
          FJ804203.1 - 2 683
          FJ804205.1 - 2 683
          FJ804206.1 - 2 683
          FJ804207.1 - 2 683
          FJ804208.1 - 2 683
          FJ804209.1 - 2 683
          FJ804210.1 - 2 683
          FJ804211.1 - 2 683
          FJ804212.1 - 2 683
          FJ804213.1 - 2 683
          FJ804214.1 - 2 683
          FJ804215.1 - 2 683
          FJ804216.1 - 2 683
          Gene Symbol: TSNAX-DISC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028393.1 6 1351
          NR_028394.1 7 1479
          NR_028395.1 7 1479
          NR_028396.1 6 1351
          NR_028397.1 6 1216
          NR_028398.1 4 970
          NR_028399.1 8 1586
          NR_028400.1 7 1458

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          neuron migration
          positive regulation of cell-matrix adhesion
          positive regulation of neuroblast proliferation
          cell proliferation in forebrain
          pyramidal neuron migration
          positive regulation of Wnt signaling pathway
          TOR signaling
          nonmotile primary cilium assembly
          positive regulation of axon extension
          mitochondrial calcium ion homeostasis
          response to electrical stimulus
          canonical Wnt signaling pathway
          regulation of dendritic spine development
          protein localization to centrosome
          regulation of synapse maturation
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          protein binding
          kinesin binding
          protein complex binding
          protein complex scaffold

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0