Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CES4A SIRNA Assays ›
Gene Symbol:
CES4A
Gene Name:
carboxylesterase 4A
Gene Aliases:
CES6, CES8
Chromosome Location:
Chr.16: 66988589 - 67009758 on Build GRCh38
UniGene ID:
Hs.346947
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS178607
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CES4A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190201.1
NP_001177130.1
5, 6
768
NM_001190202.1
NP_001177131.1
5, 6
780
NM_001318506.1
NP_001305435.1
5, 6
824
NM_173815.6
NP_776176.5
7, 8
1087
XM_005255896.4
XP_005255953.1
7, 8
1084
XM_011523021.2
XP_011521323.1
8, 9
1216
XM_011523023.2
XP_011521325.1
8, 9
1204
XM_011523024.2
XP_011521326.1
8, 9
1216
XM_011523025.2
XP_011521327.1
8, 9
1216
XM_011523026.2
XP_011521328.1
7, 8
1096
XM_011523027.2
XP_011521329.1
8, 9
1216
XM_011523028.2
XP_011521330.1
7, 8
1104
XM_011523029.2
XP_011521331.1
8, 9
1216
XM_011523030.2
XP_011521332.1
7, 8
1084
XM_017023160.1
XP_016878649.1
8, 9
1204
XM_017023161.1
XP_016878650.1
8, 9
1204
XM_017023162.1
XP_016878651.1
8, 9
1216
XM_017023163.1
XP_016878652.1
8, 9
1216
XM_017023164.1
XP_016878653.1
8, 9
1204
XM_017023165.1
XP_016878654.1
8, 9
1204
XM_017023166.1
XP_016878655.1
8, 9
1216
XM_017023167.1
XP_016878656.1
5, 6
1168
XM_017023168.1
XP_016878657.1
5, 6
1174
GenBank mRNA
AK094783.1
-
5, 6
785
AK293061.1
-
5, 6
616
AK295483.1
-
5, 6
720
AK296064.1
-
7, 8
1007
AK300792.1
-
5, 6
728
BC064573.1
-
5, 6
548
BC084555.1
-
6, 7
792
Back To Top
More Information
Panther Classification:
Molecular Function -
esterase
Gene Ontology Categories:
Process(es)
carboxylic ester hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0