Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENDOV SIRNA Assays ›
Gene Symbol:
ENDOV
Gene Name:
endonuclease V
Gene Aliases:
-
Chromosome Location:
Chr.17: 80415117 - 80438086 on Build GRCh38
UniGene ID:
Hs.389678
Species:
Human
siRNA ID
HSS178657
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ENDOV
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164637.2
NP_001158109.1
4
319
NM_001164638.1
NP_001158110.1
4
319
NM_173627.4
NP_775898.2
5
454
XM_005257244.3
XP_005257301.1
5
504
XM_005257246.3
XP_005257303.1
4
321
XM_006721837.3
XP_006721900.1
5
504
XM_011524655.1
XP_011522957.1
5
504
XM_011524656.1
XP_011522958.1
5
504
XM_011524657.1
XP_011522959.1
5
504
XM_011524658.2
XP_011522960.1
5
504
XM_011524660.1
XP_011522962.1
4
319
XM_011524661.2
XP_011522963.1
4
321
XM_011524662.1
XP_011522964.1
5
504
XM_011524663.1
XP_011522965.1
5
504
XM_011524664.2
XP_011522966.1
6
1122
XM_011524666.1
XP_011522968.1
5
686
XM_011524667.2
XP_011522969.1
5
632
XM_011524669.2
XP_011522971.1
5
504
XM_011524670.1
XP_011522972.1
5
504
XM_011524671.2
XP_011522973.1
5
504
XM_011524672.1
XP_011522974.1
4
302
XM_011524673.1
XP_011522975.1
3
259
XM_011524674.1
XP_011522976.1
4
302
XM_011524675.1
XP_011522977.1
3
279
XM_011524676.1
XP_011522978.1
4
551
XM_011524677.1
XP_011522979.1
3
262
XM_011524678.2
XP_011522980.1
3
417
XM_017024508.1
XP_016879997.1
5
1451
XM_017024509.1
XP_016879998.1
5
1397
XM_017024510.1
XP_016879999.1
5
504
XM_017024511.1
XP_016880000.1
4
321
XM_017024512.1
XP_016880001.1
5
632
XM_017024513.1
XP_016880002.1
2
377
XM_017024514.1
XP_016880003.1
4
302
XM_017024515.1
XP_016880004.1
3
262
XM_017024516.1
XP_016880005.1
2
165
XM_017024517.1
XP_016880006.1
3
279
XM_017024518.1
XP_016880007.1
4
302
XM_017024519.1
XP_016880008.1
3
262
XM_017024520.1
XP_016880009.1
2
588
GenBank mRNA
AK092539.1
-
4
319
AK096344.1
-
5
454
AK096802.1
-
3
187
AK123689.1
-
3
262
BC037889.2
-
2
176
BC045824.1
-
2
176
BC064545.1
-
4
302
5736957
BX647411.1
-
2
182
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
RNA phosphodiester bond hydrolysis, endonucleolytic
magnesium ion binding
single-stranded RNA binding
endodeoxyribonuclease activity, producing 5'-phosphomonoesters
endoribonuclease activity, producing 5'-phosphomonoesters
structure-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0