Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198477.2 | NP_940879.1 | 3 | 420 | ||
NR_133910.1 | 3 | 437 | ||||
GenBank mRNA | AK292985.1 | - | 3 | 318 | ||
AY358433.1 | - | 3 | 356 | |||
AY598464.1 | - | 3 | 420 | |||
BC093946.1 | - | 3 | 271 | 7939791 | ||
BC112095.1 | - | 3 | 273 | 8327563 | ||
BM978124.1 | - | 2 | 511 | |||
BU684368.1 | - | 2 | 515 |