Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD34A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039888.3 | NP_001034977.1 | 4 | 1763 | ||
GenBank mRNA | AK094282.1 | - | 2 | 664 | ||
AK128050.1 | - | 4 | 1757 |