Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK1 SIRNA Assays ›
Gene Symbol:
ANK1
Gene Name:
ankyrin 1
Gene Aliases:
ANK, SPH1, SPH2
Chromosome Location:
Chr.8: 41653225 - 41896762 on Build GRCh38
UniGene ID:
Hs.654438
Species:
Human
siRNA ID
HSS178893
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000037.3
NP_000028.3
20
2334
NM_001142446.1
NP_001135918.1
20
2631
NM_020475.2
NP_065208.2
20
2334
NM_020476.2
NP_065209.2
20
2334
NM_020477.2
NP_065210.2
20
2334
XM_005273476.4
XP_005273533.1
20
2608
XM_011544490.2
XP_011542792.1
21
2530
XM_011544491.2
XP_011542793.1
21
2529
XM_011544494.2
XP_011542796.1
21
2529
XM_011544495.2
XP_011542797.1
21
2528
XM_011544496.2
XP_011542798.1
21
2530
XM_011544500.2
XP_011542802.1
20
2436
XM_011544501.2
XP_011542803.1
20
2436
XM_011544502.2
XP_011542804.1
20
2436
XM_011544503.2
XP_011542805.1
21
2527
XM_011544504.2
XP_011542806.1
20
2436
XM_011544505.2
XP_011542807.1
20
2436
XM_017013319.1
XP_016868808.1
21
2529
XM_017013320.1
XP_016868809.1
21
2531
XM_017013321.1
XP_016868810.1
21
2474
XM_017013322.1
XP_016868811.1
20
2436
XM_017013323.1
XP_016868812.1
21
2450
XM_017013324.1
XP_016868813.1
20
2608
XM_017013325.1
XP_016868814.1
20
2375
XM_017013326.1
XP_016868815.1
20
2368
XM_017013327.1
XP_016868816.1
21
2529
XM_017013328.1
XP_016868817.1
21
2529
XM_017013329.1
XP_016868818.1
20
2606
GenBank mRNA
AB209418.1
-
20
2631
AK126647.1
-
3
190
M28880.1
-
20
2334
X16609.1
-
20
2334
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
signal transduction
positive regulation of organelle organization
maintenance of epithelial cell apical/basal polarity
protein targeting to plasma membrane
structural molecule activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
spectrin binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0