Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANXA2 SIRNA Assays ›
          Gene Symbol:
          ANXA2 ANXA2P2
          Gene Name:
          annexin A2
          annexin A2 pseudogene 2
          Gene Aliases:
          ANX2, ANX2L4, CAL1H, HEL-S-270, LIP2, LPC2, LPC2D, P36, PAP-IV
          ANX2L2, ANX2P2, LPC2B
          Chromosome Location:
          Chr.15: 60347151 - 60398025 on Build GRCh38
          Chromosome Location:
          Chr.9: 33624225 - 33625534 on Build GRCh38
          UniGene ID:
          Hs.511605
          Hs.534301
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS179172
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANXA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001002857.1 NP_001002857.1 14 1120
          NM_001002858.2 NP_001002858.1 13 1111
          NM_001136015.2 NP_001129487.1 14 1144
          NM_004039.2 NP_004030.1 13 1039
          XM_011521477.2 XP_011519779.1 12 1000
          XM_017022090.1 XP_016877579.1 16 4636
          XM_017022091.1 XP_016877580.1 16 4611
          XM_017022092.1 XP_016877581.1 13 1059
          XM_017022093.1 XP_016877582.1 12 939
          GenBank mRNA AK092006.1 - 10 3199
          AK124427.1 - 12 2008
          AK297923.1 - 8 604
          AK309356.1 - 1514
          BC001388.2 - 14 1108 3051155
          BC001748.1 - 13 1194
          BC009564.1 - 13 1021 3901641
          BC013843.1 - 13 1032
          BC015834.1 - 13 1043 4094952
          BC016774.1 - 13 1045 4096060
          BC021114.1 - 13 1021 5092810
          BC023990.1 - 14 1124 3961419
          BC052558.1 - 12 995 6247665
          BC052567.1 - 13 1026 6483607
          BC066955.1 - 13 1039
          BC068065.1 - 13 1019 6265952
          BC093056.1 - 14 1144 30403089
          BT007432.1 - 12 984
          BX640598.1 - 13 1110
          D00017.1 - 13 1033
          FJ224302.1 - 14 1124
          Gene Symbol: ANXA2P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003573.1 1 1033
          GenBank mRNA M62898.1 - 1 1033

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calcium-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          membrane raft assembly
          positive regulation of protein phosphorylation
          negative regulation of receptor internalization
          membrane budding
          body fluid secretion
          collagen fibril organization
          positive regulation of vesicle fusion
          negative regulation of low-density lipoprotein particle receptor catabolic process
          osteoclast development
          fibrinolysis
          negative regulation of catalytic activity
          positive regulation of fibroblast proliferation
          positive regulation of binding
          protein heterotetramerization
          protein targeting to plasma membrane
          response to thyroid hormone
          cell-cell adhesion
          negative regulation of receptor binding
          positive regulation of receptor activity
          protease binding
          calcium ion binding
          protein binding
          calcium-dependent phospholipid binding
          phosphatidylinositol-4,5-bisphosphate binding
          cytoskeletal protein binding
          Rab GTPase binding
          phospholipase A2 inhibitor activity
          receptor activator activity
          S100 protein binding
          poly(A) RNA binding
          calcium-dependent protein binding
          cadherin binding involved in cell-cell adhesion
          phospholipase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0