Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST6GALNAC6 SIRNA Assays ›
Gene Symbol:
ST6GALNAC6
Gene Name:
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
Gene Aliases:
SIAT7-F, SIAT7F, ST6GALNACVI
Chromosome Location:
Chr.9: 127885321 - 127906725 on Build GRCh38
UniGene ID:
Hs.109672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS179207
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST6GALNAC6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286999.1
NP_001273928.1
3
279
NM_001287000.1
NP_001273929.1
2
224
NM_001287001.1
NP_001273930.1
2
249
NM_001287002.1
NP_001273931.1
2
220
NM_001287003.1
NP_001273932.1
2
249
NM_013443.4
NP_038471.2
3
279
NR_104629.1
2
220
XM_011518610.2
XP_011516912.1
4
1020
XM_011518611.2
XP_011516913.1
3
965
XM_017014658.1
XP_016870147.1
2
869
XM_017014659.1
XP_016870148.1
4
1020
XM_017014660.1
XP_016870149.1
2
869
XM_017014661.1
XP_016870150.1
3
965
XM_017014662.1
XP_016870151.1
2
199
XM_017014663.1
XP_016870152.1
3
359
XM_017014664.1
XP_016870153.1
2
249
XM_017014665.1
XP_016870154.1
3
286
XM_017014666.1
XP_016870155.1
3
359
XM_017014667.1
XP_016870156.1
2
630
GenBank mRNA
AB035173.1
-
1
77
AJ507293.1
-
1
16
AK023900.1
-
2
175
AK057100.1
-
2
249
AK226174.1
-
2
105
AK300534.1
-
2
131
AK310850.1
-
2
220
AY358672.1
-
2
185
BC006564.2
-
3
196
3009935
BC007802.2
-
3
223
4128833
BC016299.2
-
3
223
3507812
BG699392.1
-
3
292
KU178617.1
-
2
104
KU178618.1
-
2
104
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
glycosylceramide metabolic process
glycosphingolipid metabolic process
glycoprotein metabolic process
oligosaccharide metabolic process
oligosaccharide biosynthetic process
cell-cell recognition
protein N-linked glycosylation via asparagine
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0