This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
ANXA6
- Gene Name:
-
annexin A6
- Gene Aliases:
-
ANX6, CBP68
- Chromosome Location:
-
Chr.5: 151100706 - 151157882 on Build GRCh38
- UniGene ID:
-
Hs.412117
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
ANXA6 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001155.4 |
NP_001146.2 |
4, 5
|
436
|
|
| NM_001193544.1 |
NP_001180473.1 |
3, 4
|
313
|
|
| XM_005268432.4 |
XP_005268489.1 |
4, 5
|
436
|
|
| GenBank mRNA |
AK126836.1 |
- |
|
301
|
|
| AK290672.1 |
- |
4, 5
|
301
|
|
| AK303078.1 |
- |
3, 4
|
299
|
|
| BC017046.1 |
- |
4, 5
|
257
|
3912460 |
| BM904172.1 |
- |
4, 5
|
542
|
|
| BQ924484.1 |
- |
4, 5
|
362
|
|
| D00510.1 |
- |
3, 4
|
208
|
|
| J03578.1 |
- |
4, 5
|
264
|
|
| KU177903.1 |
- |
3, 4
|
208
|
|
| KU177904.1 |
- |
3, 4
|
208
|
|
| KU177905.1 |
- |
3, 4
|
208
|
|
| Y00097.1 |
- |
4, 5
|
308
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System