Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-DPB1 SIRNA Assays ›
          Gene Symbol:
          HLA-DPB1 HLA-DPA1
          Gene Name:
          major histocompatibility complex, class II, DP beta 1
          major histocompatibility complex, class II, DP alpha 1
          Gene Aliases:
          DPB1, HLA-DP, HLA-DP1B, HLA-DPB
          DP(W3), DP(W4), HLA-DP1A, HLADP, HLASB, PLT1
          Chromosome Location:
          Chr.6: 33075926 - 33089696 on Build GRCh38
          Chromosome Location:
          Chr.6: 33064569 - 33080778 on Build GRCh38
          UniGene ID:
          Hs.485130
          Hs.347270
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS179250
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-DPB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002121.5 NP_002112.3 2 277
          GenBank mRNA AB208978.1 - 2 193
          AB936562.1 - 1 61
          AK291986.1 - 2 254
          AK301322.1 - 2 238
          AK303153.1 - 2 245
          AK313305.1 - 2 215
          AY656678.1 - 2 161
          AY656679.1 - 2 161
          AY656680.1 - 2 161
          AY804132.1 - 2 208
          AY804133.1 - 2 170
          AY804134.1 - 2 198
          AY804135.1 - 2 198
          AY804136.1 - 2 164
          AY804137.1 - 2 198
          AY804138.1 - 2 198
          AY804139.1 - 2 198
          AY804140.1 - 2 199
          AY804141.1 - 2 184
          AY804142.1 - 2 208
          AY804143.1 - 2 197
          AY831401.1 - 2 209
          AY831402.1 - 2 209
          AY831403.1 - 2 209
          AY831404.1 - 2 161
          BC007963.2 - 2 194 4309717
          BC013184.1 - 2 212 4341117
          BC015000.1 - 2 194 4851472
          BP213121.1 - 2 277
          DQ284441.1 - 2 196
          FR775797.1 - 2 161
          FR775798.1 - 1 61
          GQ891549.1 - 2 161
          HE603611.1 - 2 161
          HE603612.1 - 2 161
          HE614584.1 - 2 161
          HE614585.1 - 2 161
          HE614586.1 - 2 161
          HE614587.1 - 2 161
          HE614588.1 - 2 161
          HE614589.1 - 2 161
          HE614590.1 - 2 161
          HE614591.1 - 2 161
          J03041.1 - 2 230
          K01615.1 - 1 59
          KJ780721.1 - 2 161
          M21886.1 - 1 74
          M28200.1 - 2 180
          M28201.1 - 1 59
          M28202.1 - 2 137
          M83664.1 - 2 220
          X00532.1 - 1 61
          X01426.1 - 2 230
          X03067.1 - 2 209
          Gene Symbol: HLA-DPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242524.1 NP_001229453.1 1 43
          NM_001242525.1 NP_001229454.1 1 43
          GenBank mRNA BI553587.1 - 1 48

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          positive regulation of interferon-gamma production
          positive regulation of T cell proliferation
          T cell receptor signaling pathway
          positive regulation of T cell activation
          interferon-gamma-mediated signaling pathway
          immune response
          cellular response to interferon-gamma
          peptide antigen binding
          MHC class II receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0