Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPK SIRNA Assays ›
          Gene Symbol:
          HNRNPK
          Gene Name:
          heterogeneous nuclear ribonucleoprotein K
          Gene Aliases:
          AUKS, CSBP, HNRPK, TUNP
          Chromosome Location:
          Chr.9: 83968083 - 83980782 on Build GRCh38
          UniGene ID:
          Hs.522257
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS179311
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNRNPK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318186.1 NP_001305115.1 14, 15 1346
          NM_001318187.1 NP_001305116.1 14, 15 1346
          NM_001318188.1 NP_001305117.1 15, 16 1418
          NM_002140.4 NP_002131.2 15, 16 1579
          NM_031262.3 NP_112552.1 15, 16 1579
          NM_031263.3 NP_112553.1 15, 16 1418
          XM_005251960.2 XP_005252017.1 15, 16 1397
          XM_005251963.3 XP_005252020.1 14, 15 1359
          XM_005251965.2 XP_005252022.1 14, 15 1317
          XM_011518616.1 XP_011516918.1 14, 15 1359
          XM_017014668.1 XP_016870157.1 15, 16 1397
          XM_017014669.1 XP_016870158.1 14, 15 1317
          GenBank mRNA AB209562.1 - 14, 15 1275
          AB451263.1 - 13, 14 1192
          AB451390.1 - 13, 14 1192
          AK096385.1 - 14, 15 1204
          AK123117.1 - 9, 10 2144
          AK291336.1 - 15, 16 1401
          AK294067.1 - 14, 15 1376
          AK300744.1 - 14, 15 1289
          AK307962.1 - 16, 17 1239
          AY911506.1 - 14, 15 1255
          BC000355.2 - 14, 15 1314 2964383
          BC014980.1 - 15, 16 1385 4906241
          BC025321.1 - 15, 16 1337
          CR456771.1 - 13, 14 1192
          S74678.1 - 14, 15 1299
          X72727.1 - 15, 16 1401

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_057141, XM_006253552, XM_006253554, XM_006253553, XM_006253555, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001301341, NM_025279, NM_001301340, NM_001301344, NM_001301345, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mRNA splicing, via spliceosome
          ovarian follicle development
          kidney development
          liver development
          transcription from RNA polymerase II promoter
          RNA processing
          acute-phase response
          signal transduction
          peripheral nervous system development
          aging
          gene expression
          regulation of low-density lipoprotein particle clearance
          response to activity
          viral process
          protein sumoylation
          cerebellum development
          hippocampus development
          cerebral cortex development
          lung development
          negative regulation of protein binding
          cellular response to insulin stimulus
          positive regulation of RNA splicing
          camera-type eye development
          negative regulation of apoptotic process
          positive regulation of low-density lipoprotein particle receptor biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of mRNA splicing, via spliceosome
          positive regulation of receptor-mediated endocytosis
          thymus development
          positive regulation of synaptic transmission
          positive regulation of dendritic spine development
          cellular response to amino acid stimulus
          cellular response to glucose stimulus
          regulation of lipid transport by positive regulation of transcription from RNA polymerase II promoter
          cellular response to rapamycin
          positive regulation of synapse maturation
          cell-cell adhesion
          positive regulation of long-term synaptic potentiation
          response to salt
          regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of dendrite extension
          cellular response to forskolin
          positive regulation of protein localization to cell surface
          negative regulation of branching morphogenesis of a nerve
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          single-stranded DNA binding
          RNA binding
          mRNA 3'-UTR binding
          protein binding
          lamin binding
          transcription factor binding
          protein domain specific binding
          pre-mRNA 3'-splice site binding
          heat shock protein binding
          actinin binding
          ribonucleoprotein complex binding
          poly(A) RNA binding
          ATPase binding
          cadherin binding involved in cell-cell adhesion
          mRNA CDS binding
          C-rich single-stranded DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4b6rj:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0