Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HOXB3 SIRNA Assays ›
Gene Symbol:
HOXB3
HOXB-AS1
Gene Name:
homeobox B3
HOXB cluster antisense RNA 1
Gene Aliases:
HOX2, HOX2G, Hox-2.7
-
Chromosome Location:
Chr.17: 48548870 - 48590369 on Build GRCh38
Chromosome Location:
Chr.17: 48544351 - 48551241 on Build GRCh38
UniGene ID:
Hs.654560
Hs.171806
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS179335
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HOXB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002146.4
NP_002137.4
4
2128
XM_005257277.3
XP_005257334.1
5
1842
XM_005257280.3
XP_005257337.1
5
1753
XM_005257282.3
XP_005257339.1
5
1318
XM_006721854.2
XP_006721917.1
5
1860
XM_011524707.2
XP_011523009.1
5
11668
XM_011524708.2
XP_011523010.1
4
11248
XM_011524710.1
XP_011523012.1
5
4068
XM_011524719.1
XP_011523021.1
4
6549
XM_011524720.2
XP_011523022.1
4
9240
XM_011524721.2
XP_011523023.1
4
4372
XM_011524726.2
XP_011523028.1
4
1326
XM_017024557.1
XP_016880046.1
5
11416
XM_017024558.1
XP_016880047.1
5
11290
XM_017024559.1
XP_016880048.1
5
11132
XM_017024560.1
XP_016880049.1
5
11376
XM_017024561.1
XP_016880050.1
5
15213
XM_017024562.1
XP_016880051.1
5
11154
XM_017024563.1
XP_016880052.1
6
13809
GenBank mRNA
AK027609.1
-
4
2166
AK291182.1
-
5
1839
AK299226.1
-
5
1186
AK299265.1
-
5
1585
AK309721.1
-
2
1006
AK315102.1
-
3
1394
AK316245.1
-
4
1361
U59298.1
-
2
2719
X16667.1
-
4
1633
Gene Symbol: HOXB-AS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_102279.1
6
533
GenBank mRNA
AI822042.1
-
4
219
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
hematopoietic progenitor cell differentiation
transcription, DNA-templated
anterior/posterior pattern specification
rhombomere development
glossopharyngeal nerve morphogenesis
thyroid gland development
embryonic skeletal system morphogenesis
regulation of neurogenesis
cartilage development
definitive hemopoiesis
face development
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0