This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
EVX2
- Gene Name:
-
even-skipped homeobox 2
- Gene Aliases:
-
EVX-2
- Chromosome Location:
-
Chr.2: 176080107 - 176083962 on Build GRCh38
- UniGene ID:
-
Hs.452341
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System