Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128928.1 | NP_001122400.1 | 7 | 1517 | ||
NM_002194.3 | NP_002185.1 | 6 | 1373 | |||
XM_005246532.1 | XP_005246589.1 | 6 | 1154 | |||
GenBank mRNA | AB451298.1 | - | 5 | 817 | ||
AK093560.1 | - | 8 | 1475 | |||
BC015496.1 | - | 5 | 883 | 3845956 | ||
BC106006.1 | - | 6 | 917 | 4472604 | ||
CR456840.1 | - | 5 | 817 | |||
CR541707.1 | - | 5 | 817 | |||
KJ535023.1 | - | 5 | 815 | |||
KJ535042.1 | - | 5 | 816 | |||
KU178146.1 | - | 5 | 817 | |||
KU178147.1 | - | 4 | 566 | |||
L08488.1 | - | 6 | 1143 |