Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004982.3 | NP_004973.1 | 3 | 840 | ||
XM_005253358.4 | XP_005253415.1 | 4 | 809 | |||
XM_017019283.1 | XP_016874772.1 | 4 | 744 | |||
XM_017019284.1 | XP_016874773.1 | 4 | 815 | |||
GenBank mRNA | AK291821.1 | - | 4 | 648 | ||
AL546327.3 | - | 3 | 840 | |||
BC000544.2 | - | 3 | 700 | 3161776 | ||
D50312.1 | - | 3 | 764 |