This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
ARHGDIB
- Gene Name:
-
Rho GDP dissociation inhibitor beta
- Gene Aliases:
-
D4, GDIA2, GDID4, LYGDI, Ly-GDI, RAP1GN1, RhoGDI2
- Chromosome Location:
-
Chr.12: 14942015 - 14961770 on Build GRCh38
- UniGene ID:
-
Hs.504877
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
ARHGDIB |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001175.6 |
NP_001166.3 |
5, 6
|
658
|
|
| NM_001321420.1 |
NP_001308349.1 |
6, 7
|
816
|
|
| NM_001321421.1 |
NP_001308350.1 |
5, 6
|
537
|
|
| NM_001321422.1 |
NP_001308351.1 |
5, 6
|
659
|
|
| NM_001321423.1 |
NP_001308352.1 |
5, 6
|
525
|
|
| NR_135637.1 |
|
3, 4
|
244
|
|
| GenBank mRNA |
AB451315.1 |
- |
4, 5
|
412
|
|
| AB451445.1 |
- |
4, 5
|
412
|
|
| AF498927.1 |
- |
4, 5
|
412
|
|
| AK311807.1 |
- |
5, 6
|
488
|
|
| AV685495.1 |
- |
5, 6
|
459
|
|
| BC009200.2 |
- |
5, 6
|
462
|
3621138 |
| BM849963.1 |
- |
2, 3
|
106
|
|
| BP365648.1 |
- |
3, 4
|
244
|
|
| BY996333.1 |
- |
5, 6
|
522
|
|
| L07916.1 |
- |
5, 6
|
486
|
|
| L20688.1 |
- |
5, 6
|
564
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System