Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPP SIRNA Assays ›
Gene Symbol:
LPP
Gene Name:
LIM domain containing preferred translocation partner in lipoma
Gene Aliases:
-
Chromosome Location:
Chr.3: 188153050 - 188890671 on Build GRCh38
UniGene ID:
Hs.720220
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS180887
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167671.2
NP_001161143.1
10
1840
NM_001167672.2
NP_001161144.1
9
1368
NM_005578.4
NP_005569.1
10
1897
XM_005247446.4
XP_005247503.1
10
1874
XM_005247450.4
XP_005247507.1
9
1843
XM_005247451.4
XP_005247508.1
10
1874
XM_005247453.2
XP_005247510.1
9
1846
XM_011512820.2
XP_011511122.1
12
2141
XM_011512823.2
XP_011511125.1
9
3956
XM_011512827.2
XP_011511129.1
10
1797
XM_011512828.2
XP_011511130.1
10
1962
XM_011512831.2
XP_011511133.1
9
1704
XM_011512833.2
XP_011511135.1
11
1931
XM_011512834.2
XP_011511136.1
11
2022
XM_011512836.2
XP_011511138.1
6
1602
XM_017006377.1
XP_016861866.1
12
2003
XM_017006378.1
XP_016861867.1
11
2019
XM_017006379.1
XP_016861868.1
11
2154
XM_017006380.1
XP_016861869.1
11
2050
XM_017006382.1
XP_016861871.1
8
1721
GenBank mRNA
AA813328.1
-
2
270
AK302959.1
-
6
1354
AK304035.1
-
9
1310
AW301088.1
-
2
286
BC130584.1
-
9
1699
40146615
BC144076.1
-
10
1756
9052596
BG192042.1
-
1
67
CR457074.1
-
8
1651
U49957.1
-
10
1897
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
biological_process
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0