Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMAD4 SIRNA Assays ›
          Gene Symbol:
          SMAD4
          Gene Name:
          SMAD family member 4
          Gene Aliases:
          DPC4, JIP, MADH4, MYHRS
          Chromosome Location:
          Chr.18: 51030213 - 51085042 on Build GRCh38
          UniGene ID:
          Hs.75862
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS180972
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMAD4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005359.5 NP_005350.1 7 1372
          GenBank mRNA AK290770.1 - 7 1034
          BC002379.2 - 8 1067 2961238
          BX647129.1 - 3 2836
          KU178168.1 - 6 834
          KU178169.1 - 6 834
          U44378.1 - 6 962

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019275, AF056002, AB010954.
          This siRNA also targets the following transcripts in Mouse: NM_008540, BC046584, GU072919, AK004804, U79748, and others.

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ovarian follicle development
          branching involved in ureteric bud morphogenesis
          response to hypoxia
          in utero embryonic development
          gastrulation with mouth forming second
          atrioventricular valve formation
          epithelial to mesenchymal transition involved in endocardial cushion formation
          left ventricular cardiac muscle tissue morphogenesis
          positive regulation of cell proliferation involved in heart valve morphogenesis
          brainstem development
          transcription from RNA polymerase II promoter
          cellular iron ion homeostasis
          transforming growth factor beta receptor signaling pathway
          SMAD protein complex assembly
          spermatogenesis
          single fertilization
          axon guidance
          endoderm development
          mesoderm development
          cell proliferation
          negative regulation of cell proliferation
          negative regulation of cardiac muscle hypertrophy
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          neural crest cell differentiation
          regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of cell growth
          BMP signaling pathway
          positive regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of BMP signaling pathway
          somite rostral/caudal axis specification
          regulation of transforming growth factor beta2 production
          positive regulation of luteinizing hormone secretion
          somatic stem cell population maintenance
          intracellular signal transduction
          atrioventricular canal development
          endothelial cell activation
          embryonic digit morphogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of follicle-stimulating hormone secretion
          developmental growth
          neuron fate commitment
          sebaceous gland development
          formation of anatomical boundary
          regulation of binding
          positive regulation of histone H3-K4 methylation
          regulation of hair follicle development
          palate development
          uterus development
          positive regulation of SMAD protein import into nucleus
          SMAD protein signal transduction
          ventricular septum morphogenesis
          negative regulation of cell death
          endocardial cell differentiation
          female gonad morphogenesis
          interleukin-6-mediated signaling pathway
          negative regulation of ERK1 and ERK2 cascade
          response to transforming growth factor beta
          cellular response to BMP stimulus
          metanephric mesenchyme morphogenesis
          nephrogenic mesenchyme morphogenesis
          seminiferous tubule development
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          positive regulation of histone H3-K9 acetylation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          transcription factor activity, protein binding
          transcription factor activity, RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          collagen binding
          transforming growth factor beta receptor, common-partner cytoplasmic mediator activity
          identical protein binding
          protein homodimerization activity
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding
          protein heterodimerization activity
          I-SMAD binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0