Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACHE SIRNA Assays ›
          Gene Symbol:
          ACHE
          Gene Name:
          acetylcholinesterase (Cartwright blood group)
          Gene Aliases:
          ACEE, ARACHE, N-ACHE, YT
          Chromosome Location:
          Chr.7: 100889994 - 100896133 on Build GRCh38
          UniGene ID:
          Hs.154495
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181183
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACHE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000665.4 NP_000656.1 2 483
          NM_001282449.1 NP_001269378.1 2 483
          NM_001302621.1 NP_001289550.1 2 486
          NM_001302622.1 NP_001289551.1 2 388
          NM_015831.2 NP_056646.1 2 432
          XM_006715995.2 XP_006716058.1 2 388
          XM_011516225.1 XP_011514527.1 1 839
          XM_011516226.1 XP_011514528.1 1 839
          XM_011516228.2 XP_011514530.1 2 391
          XM_011516229.2 XP_011514531.1 2 332
          XM_017012219.1 XP_016867708.1 1 839
          XM_017012220.1 XP_016867709.1 1 839
          GenBank mRNA AF334270.1 - 2 398
          AK223443.1 - 2 397
          AK291321.1 - 2 383
          BC026315.1 - 2 400
          BC036813.1 - 2 401
          BC105060.1 - 2 376 8144063
          BC105062.1 - 2 376 8144065
          BC143469.1 - 2 376 9051977
          BG707892.1 - 2 492
          DA205851.1 - 2 389
          M55040.1 - 1 449

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          acetylcholine catabolic process in synaptic cleft
          regulation of receptor recycling
          osteoblast development
          DNA replication
          acetylcholine catabolic process
          phosphatidylcholine biosynthetic process
          cell adhesion
          nervous system development
          synapse assembly
          muscle organ development
          cell proliferation
          response to wounding
          receptor internalization
          negative regulation of synaptic transmission, cholinergic
          neurotransmitter biosynthetic process
          amyloid precursor protein metabolic process
          neurotransmitter receptor biosynthetic process
          positive regulation of protein secretion
          protein tetramerization
          retina development in camera-type eye
          beta-amyloid binding
          acetylcholinesterase activity
          cholinesterase activity
          protein binding
          collagen binding
          hydrolase activity
          serine hydrolase activity
          acetylcholine binding
          protein homodimerization activity
          laminin binding
          protein self-association

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0