Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CITED1 SIRNA Assays ›
          Gene Symbol:
          CITED1
          Gene Name:
          Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1
          Gene Aliases:
          MSG1
          Chromosome Location:
          Chr.X: 72301638 - 72307187 on Build GRCh38
          UniGene ID:
          Hs.40403
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181342
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CITED1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144885.1 NP_001138357.1 4 710
          NM_001144886.1 NP_001138358.1 3 662
          NM_001144887.1 NP_001138359.1 3 589
          NM_004143.3 NP_004134.2 3 1010
          XM_011530958.1 XP_011529260.1 4 701
          GenBank mRNA AB451286.1 - 2 434
          AB451418.1 - 2 434
          AI217516.1 - 1 238
          BC004240.1 - 3 963 3627230
          U65092.1 - 3 629

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          vasculogenesis
          metanephros development
          branching involved in ureteric bud morphogenesis
          placenta development
          negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          nucleocytoplasmic transport
          apoptotic process
          transforming growth factor beta receptor signaling pathway
          brain development
          cell proliferation
          positive regulation of gene expression
          negative regulation of Wnt signaling pathway
          melanocyte differentiation
          positive regulation of transforming growth factor beta receptor signaling pathway
          response to lipopolysaccharide
          response to insulin
          response to cytokine
          response to interferon-gamma
          pigmentation
          negative regulation of neuron apoptotic process
          response to estrogen
          negative regulation of osteoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          response to cAMP
          mesenchymal to epithelial transition
          SMAD protein signal transduction
          labyrinthine layer development
          spongiotrophoblast layer development
          response to interleukin-1
          response to interleukin-2
          response to interleukin-4
          response to interleukin-6
          response to interleukin-9
          response to interleukin-11
          response to parathyroid hormone
          response to transforming growth factor beta
          RNA polymerase II transcription coactivator activity
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          protein binding
          protein C-terminus binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          LBD domain binding
          co-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0