Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS181369
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
3
291
NM_001018017.2
NP_001018017.1
3
264
NM_001044390.2
NP_001037855.1
3
245
NM_001044391.2
NP_001037856.1
3
264
NM_001044392.2
NP_001037857.1
3
291
NM_001044393.2
NP_001037858.1
3
245
NM_001204285.1
NP_001191214.1
3
924
NM_001204286.1
NP_001191215.1
3
951
NM_001204287.1
NP_001191216.1
3
345
NM_001204288.1
NP_001191217.1
3
291
NM_001204292.1
NP_001191221.1
3
272
NM_001204293.1
NP_001191222.1
3
318
NM_001204294.1
NP_001191223.1
3
245
NM_001204295.1
NP_001191224.1
3
291
NM_001204296.1
NP_001191225.1
3
272
NM_001204297.1
NP_001191226.1
3
345
NM_002456.5
NP_002447.4
3
318
GenBank mRNA
AF125525.1
-
3
183
AF348143.1
-
3
270
AK301575.1
-
4
1176
AY327582.1
-
3
197
AY327583.1
-
3
216
AY327584.1
-
3
243
AY327587.1
-
3
269
AY327590.1
-
3
197
AY327591.1
-
3
197
AY327592.1
-
3
216
AY327595.1
-
3
216
AY327596.1
-
4
399
AY327597.1
-
3
197
AY327598.1
-
3
197
AY327599.1
-
3
224
AY327600.1
-
3
216
AY466157.1
-
3
173
BC120974.2
-
3
241
40115473
BC120975.2
-
3
268
40115475
EF583642.1
-
3
594
EF583643.1
-
3
422
EF583644.1
-
3
408
EF583645.1
-
3
368
EF583647.1
-
3
323
EF583648.1
-
3
250
EF583649.1
-
3
269
EF583650.1
-
3
206
EF583651.1
-
3
206
EF583655.1
-
3
269
EF670706.1
-
3
618
EF670707.1
-
3
287
EF670709.1
-
3
250
EF670710.1
-
3
242
EF670711.1
-
3
296
EF670712.1
-
3
223
EF670713.1
-
3
269
EF670716.1
-
3
269
EF670717.1
-
3
269
EF670718.1
-
3
323
EF670719.1
-
3
293
EF670721.1
-
3
250
FJ226028.1
-
2
725
FJ226029.1
-
2
883
FJ226030.1
-
3
853
FJ226031.1
-
2
811
FJ226032.1
-
2
744
FJ226033.1
-
2
725
FJ226034.1
-
3
644
FJ226036.1
-
2
744
FJ226037.1
-
3
422
FJ226038.1
-
2
743
FJ226040.1
-
3
269
FJ226042.1
-
3
323
FJ226043.1
-
3
309
FJ226044.1
-
3
349
FJ226047.1
-
3
223
FJ226049.1
-
3
323
FJ226050.1
-
3
296
FJ226052.1
-
3
242
FJ226053.1
-
3
250
FJ226054.1
-
3
368
FJ226055.1
-
3
250
J05581.1
-
3
924
J05582.1
-
3265
M34088.1
-
1
411
M34089.1
-
1
411
U60259.1
-
3
198
U60260.1
-
3
217
U60261.1
-
3
271
X52228.1
-
4
1029
X52229.1
-
4
1029
X80761.1
-
3
192
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0