Search
Search

NRDC| Gene Symbol: NRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101662.1 | NP_001095132.1 | 25 | 3109 | ||
| NM_001242361.1 | NP_001229290.1 | 27 | 2708 | |||
| NM_002525.2 | NP_002516.2 | 27 | 3313 | |||
| XM_005270903.1 | XP_005270960.1 | 25 | 2456 | |||
| XM_011541522.1 | XP_011539824.1 | 28 | 3302 | |||
| XM_011541523.1 | XP_011539825.1 | 25 | 2673 | |||
| XM_011541525.2 | XP_011539827.1 | 27 | 2771 | |||
| XM_017001375.1 | XP_016856864.1 | 25 | 2533 | |||
| GenBank mRNA | AK055516.1 | - | 25 | 2453 | ||
| AK299271.1 | - | 25 | 2556 | |||
| AK312318.1 | - | 25 | 2921 | |||
| AY049784.1 | - | 25 | 2825 | |||
| AY360265.1 | - | 27 | 2684 | |||
| BC008775.2 | - | 25 | 2883 | 3138712 | ||
| U64898.1 | - | 25 | 2881 | |||
| X93207.1 | - | 27 | 3126 | |||
| X93209.1 | - | 25 | 2922 | |||