Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015718.2 | NP_056533.1 | 4 | 376 | ||
GenBank mRNA | AF190122.1 | - | 4 | 376 | ||
AF229177.1 | - | 2 | 115 |