Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177948.1 | NP_001171419.1 | 5 | 394 | ||
NM_001177949.1 | NP_001171420.1 | 5 | 409 | |||
NM_153694.4 | NP_710161.1 | 5 | 405 | |||
XM_005268922.4 | XP_005268979.3 | 5 | 655 | |||
XM_005268924.1 | XP_005268981.1 | 5 | 393 | |||
XM_005268925.1 | XP_005268982.1 | 5 | 398 | |||
XM_005268926.3 | XP_005268983.1 | 5 | 377 | |||
XM_005268927.2 | XP_005268984.1 | 5 | 366 | |||
XM_011538421.2 | XP_011536723.2 | 5 | 577 | |||
XM_017019368.1 | XP_016874857.1 | 5 | 652 | |||
GenBank mRNA | AF492003.1 | - | 5 | 350 | ||
AF517774.1 | - | 5 | 348 | |||
AK125930.1 | - | 3 | 1060 | |||
BC062662.1 | - | 5 | 400 | 6616222 | ||
BQ428625.1 | - | 5 | 410 |