Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAX5 SIRNA Assays ›
Gene Symbol:
PAX5
Gene Name:
paired box 5
Gene Aliases:
ALL3, BSAP
Chromosome Location:
Chr.9: 36833274 - 37035949 on Build GRCh38
UniGene ID:
Hs.654464
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS181754
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280547.1
NP_001267476.1
6
1188
NM_001280548.1
NP_001267477.1
6
1188
NM_001280549.1
NP_001267478.1
6
1188
NM_001280550.1
NP_001267479.1
6
1188
NM_001280551.1
NP_001267480.1
5
1022
NM_001280552.1
NP_001267481.1
6
1188
NM_001280553.1
NP_001267482.1
5
1059
NM_001280554.1
NP_001267483.1
5
1059
NM_001280555.1
NP_001267484.1
5
990
NM_001280556.1
NP_001267485.1
5
1022
NM_016734.2
NP_057953.1
6
1188
NR_103999.1
6
1188
NR_104000.1
6
1188
XM_005251481.4
XP_005251538.1
6
1809
XM_011517896.2
XP_011516198.1
6
2658
XM_011517897.2
XP_011516199.1
6
1813
GenBank mRNA
AY463952.1
-
6
750
AY463953.1
-
6
750
AY463954.1
-
6
750
AY463955.1
-
6
750
AY463956.1
-
6
750
AY463957.1
-
6
750
FJ626406.1
-
2
164
FJ626407.1
-
2
164
FJ626408.1
-
2
164
FJ626409.1
-
2
164
FJ626410.1
-
2
164
FJ626411.1
-
2
164
FJ626412.1
-
2
164
FJ626419.1
-
5
574
FJ626420.1
-
5
574
FJ626421.1
-
6
740
FJ626422.1
-
5
611
FJ626423.1
-
5
611
FJ626424.1
-
5
542
FJ626425.1
-
6
740
M96944.1
-
6
815
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109261.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
humoral immune response
multicellular organism development
spermatogenesis
aging
organ morphogenesis
lateral ventricle development
cerebral cortex development
adult behavior
skeletal muscle cell differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic cranial skeleton morphogenesis
negative regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8hwsw:80/100.66.78.82:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0