Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SSUH2 SIRNA Assays ›
          Gene Symbol:
          SSUH2
          Gene Name:
          ssu-2 homolog (C. elegans)
          Gene Aliases:
          C3orf32, SSU-2, fls485
          Chromosome Location:
          Chr.3: 8619400 - 8745035 on Build GRCh38
          UniGene ID:
          Hs.736935
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181832
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SSUH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015931.2 NP_057015.1 4 329
          XM_011533774.1 XP_011532076.1 12 1878
          XM_017006510.1 XP_016861999.1 5 4643
          XM_017006511.1 XP_016862000.1 11 1566
          XM_017006512.1 XP_016862001.1 11 1735
          XM_017006513.1 XP_016862002.1 10 1571
          XM_017006514.1 XP_016862003.1 10 1477
          XM_017006515.1 XP_016862004.1 12 2632
          XM_017006516.1 XP_016862005.1 7 993
          XM_017006517.1 XP_016862006.1 7 946
          XM_017006518.1 XP_016862007.1 12 1857
          XM_017006519.1 XP_016862008.1 5 4588
          XM_017006520.1 XP_016862009.1 5 939
          XM_017006521.1 XP_016862010.1 8 1097
          XM_017006522.1 XP_016862011.1 8 988
          XM_017006523.1 XP_016862012.1 6 691
          XM_017006524.1 XP_016862013.1 7 929
          XM_017006525.1 XP_016862014.1 10 1498
          XM_017006526.1 XP_016862015.1 5 577
          XM_017006527.1 XP_016862016.1 11 1620
          XM_017006528.1 XP_016862017.1 5 616
          XM_017006529.1 XP_016862018.1 5 646
          XM_017006530.1 XP_016862019.1 7 1079
          XM_017006531.1 XP_016862020.1 6 1001
          XM_017006532.1 XP_016862021.1 5 5501
          XM_017006533.1 XP_016862022.1 11 1653
          XM_017006534.1 XP_016862023.1 11 1599
          XM_017006535.1 XP_016862024.1 5 5777
          XM_017006536.1 XP_016862025.1 5 5780
          XM_017006537.1 XP_016862026.1 5 5780
          XM_017006538.1 XP_016862027.1 9 1196
          XM_017006539.1 XP_016862028.1 4 308
          XM_017006540.1 XP_016862029.1 5 487
          XM_017006541.1 XP_016862030.1 2 129
          XM_017006542.1 XP_016862031.1 2 107
          GenBank mRNA AB024705.1 - 4 302
          AK093066.1 - 10 1359

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pf6sj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0