Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201965.1 | NP_001188894.1 | 5 | 1110 | ||
NM_016115.4 | NP_057199.1 | 6 | 944 | |||
NM_145863.2 | NP_665862.1 | 5 | 735 | |||
GenBank mRNA | AB209795.1 | - | 6 | 836 | ||
AF156778.1 | - | 5 | 742 | |||
AK000985.1 | - | 6 | 870 | |||
AK001283.1 | - | 5 | 668 | |||
AK056069.1 | - | 5 | 1112 | |||
AK222595.1 | - | 5 | 754 | |||
AK222715.1 | - | 5 | 726 | |||
AK303133.1 | - | 4 | 621 | |||
BC006488.2 | - | 5 | 756 | 3537359 | ||
BC009569.1 | - | 6 | 802 | 4052577 |
Gene Symbol: GPR75-ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164165.1 | NP_001157637.1 | 6 | 961 | ||
GenBank mRNA | BC110915.1 | - | 6 | 883 | 5764788 |