Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016326.3 | NP_057410.1 | 2 | 282 | ||
NM_016951.3 | NP_058647.1 | 4 | 537 | |||
NM_181640.2 | NP_857591.1 | 3 | 378 | |||
NM_181641.2 | NP_857592.1 | 3 | 441 | |||
GenBank mRNA | AF057306.1 | - | 4 | 515 | ||
AF096895.2 | - | 3 | 372 | |||
AF135380.2 | - | 4 | 531 | |||
AF135381.2 | - | 2 | 276 | |||
AF145216.2 | - | 3 | 435 | |||
AF151058.1 | - | 4 | 534 | |||
AK290307.1 | - | 3 | 352 | |||
AK301240.1 | - | 4, 5 | 528 | |||
AY358999.1 | - | 4 | 514 | |||
BC004380.1 | - | 4 | 520 | 3639550 | ||
BC091478.1 | - | 3 | 365 | 30333801 | ||
BU189908.1 | - | 3 | 338 |