Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016561.2 | NP_057645.1 | 2 | 360 | ||
XM_005255350.2 | XP_005255407.1 | 2 | 340 | |||
XM_011522520.1 | XP_011520822.1 | 2 | 340 | |||
GenBank mRNA | AF173003.1 | - | 2 | 232 | ||
AK291114.1 | - | 2 | 168 | |||
AK297067.1 | - | 2 | 168 | |||
AK300778.1 | - | 1 | 138 | |||
AK313890.1 | - | 1 | 168 | |||
BC003054.1 | - | 2 | 191 | 3537783 | ||
BI912683.1 | - | 2 | 360 |