Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282396.1 | NP_001269325.1 | 5 | 746 | ||
NM_001282397.1 | NP_001269326.1 | 3 | 424 | |||
NM_016257.3 | NP_057341.1 | 4 | 865 | |||
XM_017001447.1 | XP_016856936.1 | 4 | 478 | |||
GenBank mRNA | AB001105.1 | - | 4 | 847 | ||
AK294412.1 | - | 5 | 561 | |||
AK294813.1 | - | 3 | 406 | |||
AK312314.1 | - | 4 | 587 | |||
AL136591.1 | - | 4 | 572 | |||
BC030827.1 | - | 4 | 569 | 4811834 | ||
DC333158.1 | - | 3 | 381 |