Search
Search

S1PR5| Gene Symbol: S1PR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166215.1 | NP_001159687.1 | 2 | 775 | ||
| NM_030760.4 | NP_110387.1 | 2 | 659 | |||
| GenBank mRNA | AF331840.1 | - | 1 | 597 | ||
| AK074661.1 | - | 2 | 714 | |||
| AK094880.1 | - | 3 | 562 | |||
| BC034703.1 | - | 2 | 645 | 4754850 | ||
| BC067781.1 | - | 2 | 660 | 5286249 | ||
Set Membership: |
|