Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POLR2H SIRNA Assays ›
          Gene Symbol:
          POLR2H
          Gene Name:
          RNA polymerase II subunit H
          Gene Aliases:
          RPABC3, RPB17, RPB8
          Chromosome Location:
          Chr.3: 184361710 - 184368595 on Build GRCh38
          UniGene ID:
          Hs.432574
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS182474
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POLR2H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278698.1 NP_001265627.1 5 964
          NM_001278699.1 NP_001265628.1 5 909
          NM_001278700.1 NP_001265629.1 5 750
          NM_001278714.1 NP_001265643.1 4 816
          NM_001278715.1 NP_001265644.1 4 825
          NM_006232.3 NP_006223.2 5 900
          XM_006713666.2 XP_006713729.1 6 1502
          XM_006713667.2 XP_006713730.1 6 1251
          XM_006713668.3 XP_006713731.1 6 1447
          XM_006713670.2 XP_006713733.1 6 1447
          XM_017006636.1 XP_016862125.1 5 1354
          XM_017006637.1 XP_016862126.1 5 1363
          GenBank mRNA BC000739.2 - 5 450 3504683
          BI603667.1 - 4 452
          BM541682.1 - 5 707
          BQ065952.1 - 4 383
          U37689.1 - 5 555
          Z49199.1 - 5 468

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          transcription-coupled nucleotide-excision repair
          transcription initiation from RNA polymerase I promoter
          transcription elongation from RNA polymerase I promoter
          termination of RNA polymerase I transcription
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          7-methylguanosine mRNA capping
          transcription from RNA polymerase III promoter
          fibroblast growth factor receptor signaling pathway
          gene expression
          gene silencing by RNA
          positive regulation of type I interferon production
          somatic stem cell population maintenance
          snRNA transcription from RNA polymerase II promoter
          positive regulation of gene expression, epigenetic
          positive regulation of viral transcription
          RNA polymerase I activity
          RNA polymerase II activity
          RNA polymerase III activity
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0