Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTRF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114184.2 | NP_001107656.1 | 4 | 668 | ||
NM_001301047.2 | NP_001287976.1 | 4 | 560 | |||
NM_001301870.1 | NP_001288799.1 | 4 | 560 | |||
NM_001301871.1 | NP_001288800.1 | 4 | 643 | |||
NM_019041.6 | NP_061914.3 | 4 | 668 | |||
NR_126056.1 | 4 | 643 | ||||
XM_017010959.1 | XP_016866448.1 | 5 | 5275 | |||
XM_017010960.1 | XP_016866449.1 | 5 | 684 | |||
GenBank mRNA | AK027684.1 | - | 4 | 513 | ||
AK095240.1 | - | 4 | 578 | |||
AK225321.1 | - | 4 | 542 | |||
AK225970.1 | - | 4 | 542 | |||
AK290662.1 | - | 4 | 542 | |||
AK297671.1 | - | 4 | 535 | |||
BC011873.2 | - | 4 | 565 | 3029407 | ||
BC014428.1 | - | 4 | 554 | 4646472 | ||
BC105973.1 | - | 4 | 459 | 6049942 | ||
BX376520.2 | - | 4 | 536 |