Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APTX SIRNA Assays ›
          Gene Symbol:
          APTX
          Gene Name:
          aprataxin
          Gene Aliases:
          AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
          Chromosome Location:
          Chr.9: 32883872 - 33025129 on Build GRCh38
          UniGene ID:
          Hs.20158
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS182673
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APTX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195248.1 NP_001182177.1 7 867
          NM_001195249.1 NP_001182178.1 7 1036
          NM_001195250.1 NP_001182179.1 6 705
          NM_001195251.1 NP_001182180.1 8 973
          NM_001195252.1 NP_001182181.1 7 651
          NM_001195254.1 NP_001182183.1 6 727
          NM_175069.2 NP_778239.1 7 867
          NM_175073.2 NP_778243.1 8 973
          NR_036576.1 7 862
          NR_036577.1 7 752
          NR_036578.1 7 884
          NR_036579.1 7 1031
          XM_006716791.3 XP_006716854.1 7 875
          XM_006716792.2 XP_006716855.1 7 893
          XM_011517938.1 XP_011516240.1 6 851
          XM_011517939.2 XP_011516241.1 6 806
          XM_017014837.1 XP_016870326.1 6 956
          XM_017014838.1 XP_016870327.1 7 849
          GenBank mRNA AB603741.1 - 6 763
          AK000164.1 - 6 870
          AK055672.1 - 7 854
          AK131046.1 - 7 819
          AY040777.1 - 7 842
          AY208829.1 - 7 842
          AY208830.1 - 6 705
          AY208831.1 - 7 617
          AY208832.1 - 6 680
          AY208833.1 - 6 680
          AY208834.1 - 6 781
          AY208835.1 - 7 822
          AY208836.1 - 7 982
          AY208837.1 - 8 939
          AY208838.1 - 6 798
          AY208839.1 - 7 976
          AY208840.1 - 7 839
          AY208841.1 - 6 676
          AY208842.1 - 4 476
          AY302067.1 - 6 985
          AY302068.1 - 6 980
          AY302069.1 - 6 870
          AY302070.1 - 6 865
          AY302071.1 - 5 758
          AY302072.1 - 5 753
          AY302073.1 - 5 643
          AY302074.1 - 5 638
          BC001628.1 - 6 690
          BC032650.1 - 7 908 5505120
          BC068107.1 - 8 948
          BC104881.1 - 7 881 8143884
          BX538161.1 - 8 952

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_148889, XM_008763607, XM_006238028, XM_006238027, XM_006238026, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001025444, NM_025545, NM_001025445, BC068309, BC071213, and others.

          Panther Classification:

          Molecular Function -

          damaged DNA-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          DNA ligation
          cellular response to DNA damage stimulus
          dephosphorylation
          regulation of protein stability
          response to hydrogen peroxide
          nucleic acid phosphodiester bond hydrolysis
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          double-stranded RNA binding
          protein binding
          phosphoglycolate phosphatase activity
          DNA 5'-adenosine monophosphate hydrolase activity
          metal ion binding
          protein N-terminus binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0