Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM208B SIRNA Assays ›
Gene Symbol:
FAM208B
Gene Name:
family with sequence similarity 208 member B
Gene Aliases:
C10orf18, bA318E3.2
Chromosome Location:
Chr.10: 5684759 - 5763740 on Build GRCh38
UniGene ID:
Hs.610717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182723
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM208B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321783.1
NP_001308712.1
12
1733
NM_001321784.1
NP_001308713.1
12
1779
NM_001321785.1
NP_001308714.1
12
1733
NM_017782.4
NP_060252.4
11
1660
XM_005252474.3
XP_005252531.1
14
8471
XM_005252475.3
XP_005252532.1
15
2132
XM_005252476.4
XP_005252533.1
13
1915
XM_005252480.4
XP_005252537.1
11
1720
XM_005252481.4
XP_005252538.1
9
1205
XM_005252482.2
XP_005252539.1
13
1565
XM_011519515.2
XP_011517817.1
15
8609
XM_011519516.2
XP_011517818.1
15
8609
XM_011519517.2
XP_011517819.1
15
2529
XM_011519518.2
XP_011517820.1
14
1842
XM_011519519.2
XP_011517821.1
15
8609
XM_011519520.2
XP_011517822.1
12
8306
XM_011519521.2
XP_011517823.1
14
2053
XM_011519522.2
XP_011517824.1
14
2437
XM_011519525.2
XP_011517827.1
12
1473
XM_017016361.1
XP_016871850.1
15
8609
XM_017016362.1
XP_016871851.1
13
2336
XM_017016363.1
XP_016871852.1
14
1994
XM_017016364.1
XP_016871853.1
12
2198
XM_017016365.1
XP_016871854.1
13
1860
GenBank mRNA
AK128211.1
-
1
593
BX649177.1
-
3
633
CR936780.1
-
11
1661
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0