Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BANP SIRNA Assays ›
Gene Symbol:
BANP
RRN3P2
Gene Name:
BTG3 associated nuclear protein
RRN3 homolog, RNA polymerase I transcription factor pseudogene 2
Gene Aliases:
BEND1, SMAR1, SMARBP1
-
Chromosome Location:
Chr.16: 87949216 - 88077318 on Build GRCh38
Chromosome Location:
Chr.16: 29074842 - 29116718 on Build GRCh38
UniGene ID:
Hs.461705
Hs.720348
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS182765
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BANP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173539.1
NP_001167010.1
4
387
NM_001173540.1
NP_001167011.1
4
405
NM_001173541.1
NP_001167012.1
4
387
NM_001173542.1
NP_001167013.1
4
426
NM_001173543.1
NP_001167014.1
4
432
NM_017869.3
NP_060339.2
4
387
NM_079837.2
NP_524576.2
4
387
XM_005256008.3
XP_005256065.1
4
403
XM_005256009.3
XP_005256066.1
4
385
XM_011523170.2
XP_011521472.1
4
403
XM_011523172.2
XP_011521474.1
5
648
XM_011523174.2
XP_011521476.1
4
1174
XM_011523175.2
XP_011521477.1
4
805
XM_011523176.2
XP_011521478.1
4
867
XM_011523177.2
XP_011521479.1
4
1375
XM_011523178.2
XP_011521480.1
4
403
XM_011523179.2
XP_011521481.1
4
385
XM_011523180.2
XP_011521482.1
5
467
XM_011523181.2
XP_011521483.1
4
403
XM_011523182.1
XP_011521484.1
4
403
XM_011523183.2
XP_011521485.1
4
380
XM_011523184.2
XP_011521486.1
4
336
XM_011523186.2
XP_011521488.1
2
165
XM_017023378.1
XP_016878867.1
5
507
XM_017023379.1
XP_016878868.1
4
787
XM_017023380.1
XP_016878869.1
4
387
XM_017023381.1
XP_016878870.1
4
405
XM_017023383.1
XP_016878872.1
4
352
GenBank mRNA
AK001039.1
-
4
387
AK094158.1
-
4
433
AK125232.1
-
4
385
AK293472.1
-
4
349
AK297945.1
-
4
359
AK315089.1
-
4
385
AK315535.1
-
4
426
BC009424.1
-
4
332
3530821
DA632793.1
-
4
387
KU178664.1
-
3
235
KU178665.1
-
3
235
KU178666.1
-
3
235
KU178667.1
-
3
235
KU178668.1
-
3
235
KU178669.1
-
3
235
KU178670.1
-
3
257
Gene Symbol: RRN3P2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_003369.2
17
1878
GenBank mRNA
AK225670.1
-
17
1728
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cell cycle
multicellular organism development
covalent chromatin modification
protein localization to nucleus
negative regulation of protein catabolic process
positive regulation of transcription, DNA-templated
regulation of signal transduction by p53 class mediator
p53 binding
DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0