Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017907.2 | NP_060377.1 | 3 | 391 | ||
GenBank mRNA | AF289583.1 | - | 3 | 282 | ||
AK000632.1 | - | 3 | 280 | |||
AK300681.1 | - | 3 | 246 | |||
BC001706.1 | - | 3 | 253 | 3544000 | ||
BX381569.2 | - | 3 | 391 | |||
CR457247.1 | - | 3 | 228 |