Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018189.3 | NP_060659.3 | 3, 4 | 395 | ||
XM_011512954.2 | XP_011511256.1 | 3, 4 | 674 | |||
GenBank mRNA | AK001575.1 | - | 3, 4 | 337 | ||
AK022698.1 | - | 3, 4 | 352 | |||
AK290992.1 | - | 3, 4 | 388 | |||
AK298623.1 | - | 3, 4 | 345 | |||
AK299290.1 | - | 3, 4 | 338 | |||
BC032846.2 | - | 3, 4 | 369 | 5267235 |