Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113378.1 | NP_001106849.1 | 11 | 986 | ||
NM_018193.2 | NP_060663.2 | 11 | 986 | |||
XM_011521756.2 | XP_011520058.1 | 11 | 1010 | |||
XM_011521757.2 | XP_011520059.1 | 11 | 1006 | |||
XM_011521764.2 | XP_011520066.1 | 11 | 1010 | |||
XM_011521767.2 | XP_011520069.1 | 11 | 980 | |||
GenBank mRNA | AK001581.1 | - | 5 | 375 | ||
AK055176.1 | - | 11 | 945 | |||
BC140769.1 | - | 11 | 943 | 9021637 | ||
BC144483.1 | - | 11 | 943 | 9053010 | ||
EF469766.1 | - | 10 | 896 | |||
EF567077.1 | - | 10 | 896 |