Search
Search

PPP2R5B| Gene Symbol: PPP2R5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006244.3 | NP_006235.1 | 2 | 784 | ||
| XM_011545132.2 | XP_011543434.1 | 3 | 217 | |||
| XM_011545133.1 | XP_011543435.1 | 4 | 337 | |||
| XM_011545134.2 | XP_011543436.1 | 2 | 1623 | |||
| GenBank mRNA | AK092730.1 | - | 2 | 768 | ||
| BC045619.1 | - | 2 | 778 | 5294756 | ||
| L42374.1 | - | 2 | 487 | |||