Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018437.4 | NP_060907.2 | 4 | 552 | ||
NM_197978.2 | NP_932095.1 | 3 | 606 | |||
XM_005252086.1 | XP_005252143.1 | 4 | 514 | |||
XM_011518845.2 | XP_011517147.1 | 4 | 426 | |||
XM_011518846.2 | XP_011517148.1 | 4 | 536 | |||
GenBank mRNA | AF130060.2 | - | 3 | 339 | ||
AF228713.2 | - | 4 | 459 | |||
AF306682.1 | - | 3 | 388 | |||
AF306683.1 | - | 3 | 354 | |||
AF322875.1 | - | 4 | 458 | |||
AK292441.1 | - | 4 | 478 | |||
AL833094.1 | - | 3 | 398 | |||
AY244805.1 | - | 3 | 552 | |||
AY255672.1 | - | 3 | 457 | |||
BC048324.1 | - | 4 | 461 | 5166612 | ||
BG571634.1 | - | 3 | 430 | |||
BG717036.1 | - | 4 | 497 |