Search
Search

DNAH3| Gene Symbol: DNAH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017539.2 | NP_060009.1 | 48 | 7515 | ||
| XM_006721062.1 | XP_006721125.1 | 48 | 7653 | |||
| XM_011545882.2 | XP_011544184.1 | 49 | 8093 | |||
| XM_011545883.1 | XP_011544185.1 | 49 | 7861 | |||
| XM_011545885.2 | XP_011544187.1 | 49 | 8093 | |||
| XM_011545886.2 | XP_011544188.1 | 41 | 6468 | |||
| XM_011545888.2 | XP_011544190.1 | 31 | 4912 | |||
| XM_011545889.2 | XP_011544191.1 | 31 | 4876 | |||
| XM_017023426.1 | XP_016878915.1 | 48 | 7958 | |||
| XM_017023427.1 | XP_016878916.1 | 47 | 7850 | |||
| XM_017023428.1 | XP_016878917.1 | 46 | 7514 | |||
| XM_017023430.1 | XP_016878919.1 | 7 | 1535 | |||
| XM_017023432.1 | XP_016878921.1 | 48 | 7653 | |||
| GenBank mRNA | AF494040.1 | - | 48 | 7515 | ||