Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEZT SIRNA Assays ›
Gene Symbol:
VEZT
Gene Name:
vezatin, adherens junctions transmembrane protein
Gene Aliases:
VEZATIN
Chromosome Location:
Chr.12: 95217746 - 95302799 on Build GRCh38
UniGene ID:
Hs.24135
Species:
Human
siRNA ID
HSS183128
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VEZT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017599.3
NP_060069.3
7
1083
NR_038241.1
8
1156
NR_038242.1
7
1091
XM_006719477.3
XP_006719540.1
7
1095
XM_006719478.2
XP_006719541.1
6
989
XM_006719479.2
XP_006719542.1
8
1166
XM_006719480.3
XP_006719543.1
8
1143
XM_006719481.2
XP_006719544.1
7
1013
XM_006719482.3
XP_006719545.1
7
1394
XM_006719483.3
XP_006719546.1
7
1411
XM_006719484.3
XP_006719547.1
6
1321
XM_006719485.2
XP_006719548.1
5
763
XM_006719486.3
XP_006719549.1
5
751
XM_006719490.3
XP_006719553.1
8
1154
XM_011538543.1
XP_011536845.1
8
1088
XM_011538544.1
XP_011536846.1
8
1076
XM_011538545.2
XP_011536847.1
7
1056
XM_011538546.1
XP_011536848.1
9
1159
XM_011538547.1
XP_011536849.1
9
1203
XM_011538548.2
XP_011536850.1
6
849
XM_011538549.1
XP_011536851.1
8
1088
XM_011538550.2
XP_011536852.1
8
1088
XM_011538551.2
XP_011536853.1
8
1076
XM_011538552.1
XP_011536854.1
8
1162
XM_011538553.2
XP_011536855.1
8
1092
XM_017019586.1
XP_016875075.1
7
1044
XM_017019587.1
XP_016875076.1
6
977
XM_017019588.1
XP_016875077.1
9
1147
XM_017019589.1
XP_016875078.1
8
1131
XM_017019590.1
XP_016875079.1
9
1191
XM_017019591.1
XP_016875080.1
7
1382
XM_017019592.1
XP_016875081.1
6
836
XM_017019593.1
XP_016875082.1
9
1211
XM_017019594.1
XP_016875083.1
8
1073
XM_017019595.1
XP_016875084.1
7
1095
XM_017019596.1
XP_016875085.1
7
1095
XM_017019597.1
XP_016875086.1
7
1083
XM_017019598.1
XP_016875087.1
7
1083
XM_017019599.1
XP_016875088.1
7
1044
XM_017019600.1
XP_016875089.1
7
1399
XM_017019601.1
XP_016875090.1
8
1150
XM_017019602.1
XP_016875091.1
9
1147
XM_017019603.1
XP_016875092.1
7
1411
XM_017019604.1
XP_016875093.1
5
763
XM_017019605.1
XP_016875094.1
5
751
GenBank mRNA
AF216644.1
-
7
1010
AF225417.1
-
7
1054
AF277625.1
-
3
522
AK001338.1
-
5
764
AK225303.1
-
8
1096
AK225356.1
-
8
1105
AK307649.1
-
8
1162
AK307864.1
-
5
750
AK309943.1
-
7
1030
AL137497.1
-
8
1078
BC028597.1
-
10
1412
BC064939.1
-
8
1073
6159768
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
single organismal cell-cell adhesion
myosin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-9jqhn:80/10.239.160.220:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0