Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL3 SIRNA Assays ›
Gene Symbol:
MBNL3
Gene Name:
muscleblind like splicing regulator 3
Gene Aliases:
CHCR, MBLX, MBLX39, MBXL
Chromosome Location:
Chr.X: 132369314 - 132490030 on Build GRCh38
UniGene ID:
Hs.105134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS183277
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170701.1
NP_001164172.1
2
161
NM_001170702.1
NP_001164173.1
2
161
NM_001170703.1
NP_001164174.1
2
94
NM_001170704.1
NP_001164175.1
3
361
NM_018388.3
NP_060858.2
2
324
NM_133486.2
NP_597846.1
2
324
XM_005262431.3
XP_005262488.1
3
1136
XM_005262434.3
XP_005262491.1
3
1131
XM_005262436.3
XP_005262493.1
2
1024
XM_005262437.3
XP_005262494.1
2
162
XM_005262438.3
XP_005262495.1
2
162
XM_005262439.3
XP_005262496.1
2
94
XM_005262440.3
XP_005262497.1
4
1181
XM_005262441.3
XP_005262498.1
2
285
XM_005262442.3
XP_005262499.1
2
256
XM_017029632.1
XP_016885121.1
3
8566
XM_017029633.1
XP_016885122.1
4
1301
XM_017029634.1
XP_016885123.1
3
8566
XM_017029635.1
XP_016885124.1
3
8566
XM_017029636.1
XP_016885125.1
3
1065
XM_017029637.1
XP_016885126.1
3
8566
XM_017029638.1
XP_016885127.1
4
1199
XM_017029639.1
XP_016885128.1
2
274
GenBank mRNA
AB077698.1
-
2
324
AB077699.1
-
2
324
AF467070.1
-
2
324
AF491305.1
-
2
245
AJ427918.1
-
2
324
AJ427919.1
-
2
324
AK002178.1
-
2
130
AK300248.1
-
2
93
AK307194.1
-
4
1178
AL133625.1
-
1
69
AY072692.1
-
2
324
AY372211.1
-
2
95
BC042090.1
-
3
360
5761647
BC074775.2
-
2
287
30915333
BC074776.2
-
2
287
30915555
BU569439.1
-
2
164
DB023330.1
-
2
94
DC380677.1
-
2
290
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
multicellular organism development
RNA splicing
regulation of RNA splicing
negative regulation of myoblast differentiation
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0