Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSNAXIP1 SIRNA Assays ›
Gene Symbol:
TSNAXIP1
Gene Name:
translin associated factor X interacting protein 1
Gene Aliases:
TXI1
Chromosome Location:
Chr.16: 67806573 - 67828068 on Build GRCh38
UniGene ID:
Hs.632212
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS183293
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSNAXIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288990.1
NP_001275919.1
12
1810
NM_001288991.1
NP_001275920.1
10
1603
NM_001288992.1
NP_001275921.1
8
1556
NM_001288993.1
NP_001275922.1
9
1683
NM_001288994.1
NP_001275923.1
7
1359
NM_018430.3
NP_060900.2
12
1999
XM_005256049.4
XP_005256106.1
12
2004
XM_011523230.2
XP_011521532.1
11
2211
XM_011523231.2
XP_011521533.1
11
2190
XM_011523232.2
XP_011521534.1
12
2163
XM_011523233.2
XP_011521535.1
12
2136
XM_011523234.2
XP_011521536.1
11
2079
XM_011523235.1
XP_011521537.1
11
1846
XM_011523236.2
XP_011521538.1
9
1469
XM_011523237.2
XP_011521539.1
9
1971
XM_011523238.1
XP_011521540.1
8
1384
XM_011523239.1
XP_011521541.1
9
1385
XM_011523240.1
XP_011521542.1
10
1571
XM_011523241.1
XP_011521543.1
9
1659
XM_011523242.1
XP_011521544.1
8
1512
XM_011523243.1
XP_011521545.1
8
1466
XM_017023445.1
XP_016878934.1
9
1448
XM_017023446.1
XP_016878935.1
9
1315
XM_017023447.1
XP_016878936.1
9
1337
XM_017023448.1
XP_016878937.1
9
1288
XM_017023449.1
XP_016878938.1
10
1262
XM_017023450.1
XP_016878939.1
11
1475
XM_017023451.1
XP_016878940.1
11
1661
XM_017023452.1
XP_016878941.1
8
1491
XM_017023453.1
XP_016878942.1
10
1563
XM_017023454.1
XP_016878943.1
9
1416
XM_017023455.1
XP_016878944.1
9
1305
GenBank mRNA
AF132730.1
-
9
1487
AK027245.1
-
12
1717
AK301920.1
-
12
1583
AK302300.1
-
992
AK303841.1
-
10
1393
BC111018.1
-
12
1753
5168384
BU607826.1
-
5
690
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
biological_process
cell differentiation
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0