Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018475.4 | NP_060945.2 | 4 | 1040 | ||
NR_073070.1 | 5 | 1143 | ||||
XM_011534394.2 | XP_011532696.1 | 4 | 1030 | |||
XM_017008412.1 | XP_016863901.1 | 6 | 958 | |||
GenBank mRNA | AF132746.1 | - | 4 | 763 | ||
AF183409.1 | - | 4 | 763 | |||
AF220188.1 | - | 4 | 957 | |||
AK026163.1 | - | 3 | 423 | |||
AK054712.1 | - | 3 | 1220 | |||
AK290663.1 | - | 4 | 993 | |||
AK295289.1 | - | 5 | 1093 | |||
BC003545.1 | - | 3 | 461 | |||
BC104978.1 | - | 4 | 838 | 8143981 | ||
BC104980.1 | - | 4 | 838 | 8143983 | ||
BC107582.1 | - | 4 | 1000 | 4691718 |