Search
Search

DBNDD2
SYS1-DBNDD2| Gene Symbol: DBNDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048221.2 | NP_001041686.1 | 3 | 394 | ||
| NM_001048222.2 | NP_001041687.1 | 3 | 394 | |||
| NM_001048223.2 | NP_001041688.1 | 3 | 343 | |||
| NM_001048224.2 | NP_001041689.1 | 3 | 343 | |||
| NM_001048225.2 | NP_001041690.2 | 2 | 669 | |||
| NM_001048226.2 | NP_001041691.2 | 2 | 669 | |||
| NM_001197139.1 | NP_001184068.1 | 3 | 227 | |||
| NM_001197140.1 | NP_001184069.1 | 3 | 273 | |||
| NM_018478.3 | NP_060948.3 | 3 | 715 | |||
| GenBank mRNA | AF220191.1 | - | 3 | 721 | ||
| AJ276469.1 | - | 3 | 312 | |||
| AL591565.1 | - | 3 | 227 | |||
| AY113697.1 | - | 2 | 321 | |||
| BC001105.2 | - | 3 | 237 | 2959575 | ||
| BC012818.2 | - | 3 | 237 | 2959575 | ||
| BG703352.1 | - | 3 | 343 | |||
| BM767886.1 | - | 3 | 343 | |||
| BP308674.1 | - | 2 | 352 | |||
| BP330715.1 | - | 3 | 394 | |||
| BX335176.2 | - | 3 | 377 | |||
| CB155210.1 | - | 3 | 273 | |||
| CN347771.1 | - | 3 | 227 | |||
| DT216122.1 | - | 2 | 383 | |||
| Gene Symbol: SYS1-DBNDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003189.2 | 5 | 578 | |||
| GenBank mRNA | BC023562.2 | - | 5 | 547 | ||
| BI199175.1 | - | 4 | 442 | |||
| DN994280.1 | - | 4 | 478 | |||