Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAP2K5 SIRNA Assays ›
          Gene Symbol:
          MAP2K5
          Gene Name:
          mitogen-activated protein kinase kinase 5
          Gene Aliases:
          HsT17454, MAPKK5, MEK5, PRKMK5
          Chromosome Location:
          Chr.15: 67542606 - 67807117 on Build GRCh38
          UniGene ID:
          Hs.114198
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS183391
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAP2K5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206804.1 NP_001193733.1 4 339
          NM_002757.3 NP_002748.1 4 923
          NM_145160.2 NP_660143.1 4 923
          XM_011521787.2 XP_011520089.1 4 1000
          XM_011521788.2 XP_011520090.1 4 1000
          XM_017022414.1 XP_016877903.1 4 1000
          GenBank mRNA AK293459.1 - 4 339
          AK315435.1 - 4 470
          BC008838.2 - 4 580 4100679
          BT006780.1 - 4 270
          CR542229.1 - 4 270
          U71087.1 - 4 331
          U71088.2 - 4 275

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017246, NM_001033987, XM_008766221, XM_006243229, BC078860, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011840, AK020716, BC028260, AK166137, BC013697, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          signal transduction
          heart development
          peptidyl-tyrosine phosphorylation
          positive regulation of cell growth
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of heterotypic cell-cell adhesion
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of interleukin-8 biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of epithelial cell proliferation
          positive regulation of protein metabolic process
          negative regulation of response to cytokine stimulus
          ERK5 cascade
          cellular response to growth factor stimulus
          cellular response to laminar fluid shear stress
          negative regulation of cell migration involved in sprouting angiogenesis
          negative regulation of chemokine (C-X-C motif) ligand 2 production
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protein kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein tyrosine kinase activity
          protein binding
          ATP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline