Search
Search

TEX12| Gene Symbol: TEX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031275.4 | NP_112565.1 | 3, 4 | 308 | ||
| GenBank mRNA | AF285600.1 | - | 2, 3 | 214 | ||
| BC029506.1 | - | 3, 4 | 308 | 5298531 | ||
| BI561514.1 | - | 3, 4 | 312 | |||